Assay Details
| Gene Symbol: CELA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174048.2 | NP_776473.1 | 7 - 8 | 791 | 66 | |
| GenBank mRNA | BC149525.1 | - | 7 - 8 | 808 | 66 | |
| M80838.1 | - | 7 - 8 | 796 | 66 | ||
More Information
Set Membership: |
|

CELA1
Genomic Map