Assay Details
| Gene Symbol: IBSP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174084.3 | NP_776509.1 | 4 - 5 | 230 | 87 | |
| GenBank mRNA | S73144.1 | - | 4 - 5 | 234 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IBSP| Gene Symbol: IBSP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174084.3 | NP_776509.1 | 4 - 5 | 230 | 87 | |
| GenBank mRNA | S73144.1 | - | 4 - 5 | 234 | 87 | |
Set Membership: |
|