Assay Details
| Gene Symbol: IL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174088.1 | NP_776513.1 | 4 - 5 | 447 | 70 | |
| GenBank mRNA | EU276074.1 | - | 4 - 5 | 447 | 70 | |
| U00799.1 | - | 4 - 5 | 447 | 70 | ||
More Information
Set Membership: |
|

IL10
Genomic Map