Assay Details
| Gene Symbol: IL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174091.2 | NP_776516.1 | 4 - 5 | 369 | 106 | |
| GenBank mRNA | AF124789.1 | - | 4 - 5 | 369 | 106 | |
| AF173175.1 | - | 3 - 4 | 175 | 106 | ||
| BC102879.1 | - | 4 - 5 | 403 | 106 | ||
| EU276078.1 | - | 3 - 4 | 220 | 106 | ||
| EU574909.1 | - | 3 - 4 | 220 | 106 | ||
More Information
Set Membership: |
|

IL18
Genomic Map