Assay Details
| Gene Symbol: LIPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174103.2 | NP_776528.1 | 9 - 10 | 1030 | 74 | |
| GenBank mRNA | L26319.1 | - | 9 - 10 | 1030 | 74 | |
More Information
Set Membership: |
|

LIPF
Genomic Map