Assay Details
| Gene Symbol: NPFF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174123.3 | NP_776548.1 | 1 - 2 | 115 | 113 | |
| GenBank mRNA | AF148699.1 | - | 1 - 2 | 116 | 113 | |
More Information
Set Membership: |
|

NPFF
Genomic Map