Assay Details
| Gene Symbol: ODC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174130.2 | NP_776555.1 | 3 - 4 | 466 | 63 | |
| GenBank mRNA | BC146218.1 | - | 4 - 5 | 577 | 63 | |
| M92441.1 | - | 3 - 4 | 466 | 63 | ||
More Information
Set Membership: |
|

ODC1
Genomic Map