Assay Details
| Gene Symbol: OLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174132.2 | NP_776557.1 | 5 - 6 | 699 | 78 | |
| GenBank mRNA | BT029784.1 | - | 5 - 6 | 703 | 78 | |
| D89049.1 | - | 5 - 6 | 699 | 78 | ||
More Information
Set Membership: |
|

OLR1
Genomic Map