Assay Details
| Gene Symbol: OXTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174134.2 | NP_776559.1 | 1 - 2 | 1081 | 74 | |
| GenBank mRNA | AF101724.1 | - | 1 - 2 | 1081 | 74 | |
More Information
Set Membership: |
|

OXTR
Genomic Map