Assay Details
| Gene Symbol: PARG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174138.3 | NP_776563.1 | 17 - 18 | 3034 | 151 | |
| GenBank mRNA | U78975.1 | - | 17 - 18 | 3037 | 151 | |
More Information
Set Membership: |
|

PARG
Genomic Map