Assay Details
| Gene Symbol: PIGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174143.1 | NP_776568.1 | 4 - 5 | 1193 | 78 | |
| GenBank mRNA | BC149032.1 | - | 3 - 4 | 1208 | 78 | |
| L04797.1 | - | 3 - 4 | 1114 | 78 | ||
| X81371.1 | - | 1193 | 78 | |||
More Information
Set Membership: |
|

PIGR
Genomic Map