Assay Details
| Gene Symbol: PIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174144.2 | NP_776569.1 | 4 - 5 | 1019 | 79 | |
| GenBank mRNA | AF259078.1 | - | 4 - 5 | 1019 | 79 | |
More Information
Set Membership: |
|

PIM1
Genomic Map