Assay Details
| Gene Symbol: PLAU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174147.2 | NP_776572.1 | 10 - 11 | 1247 | 77 | |
| GenBank mRNA | BC122657.1 | - | 10 - 11 | 1194 | 77 | |
| BT030737.1 | - | 10 - 11 | 1241 | 77 | ||
| L03546.1 | - | 10 - 11 | 1247 | 77 | ||
| X85801.1 | - | 1099 | 77 | |||
More Information
Set Membership: |
|

PLAU
Genomic Map