Assay Details
| Gene Symbol: SARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174175.3 | NP_776600.1 | 3 - 4 | 365 | 114 | |
| GenBank mRNA | AF297553.1 | - | 3 - 4 | 297 | 114 | |
More Information
Set Membership: |
|

SARS1
Genomic Map