Assay Details
| Gene Symbol: SARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174175.3 | NP_776600.1 | 4 - 5 | 536 | 65 | |
| GenBank mRNA | AF297553.1 | - | 4 - 5 | 468 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SARS1| Gene Symbol: SARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174175.3 | NP_776600.1 | 4 - 5 | 536 | 65 | |
| GenBank mRNA | AF297553.1 | - | 4 - 5 | 468 | 65 | |
Set Membership: |
|