Assay Details
| Gene Symbol: SCG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174176.3 | NP_776601.1 | 1 - 2 | 78 | 85 | |
| GenBank mRNA | BC103034.1 | - | 1 - 2 | 79 | 85 | |
| J05468.1 | - | 1 - 2 | 79 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103034.1, NM_174176.3 transcripts.
Set Membership: |
|

SCG2
Genomic Map