Assay Details
| Gene Symbol: SDHD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174179.2 | NP_776604.1 | 2 - 3 | 174 | 85 | |
| GenBank mRNA | AY770191.1 | - | 2 - 3 | 160 | 85 | |
| BC102257.1 | - | 2 - 3 | 194 | 85 | ||
| FJ495085.1 | - | 2 - 3 | 191 | 85 | ||
| U50987.1 | - | 2 - 3 | 174 | 85 | ||
More Information
Set Membership: |
|

SDHD
Genomic Map