Assay Details
| Gene Symbol: WARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174218.1 | NP_776643.1 | 1 - 2 | 293 | 63 | |
| GenBank mRNA | BC102806.1 | - | 2 - 3 | 238 | 63 | |
| X52113.1 | - | 220 | 63 | |||
| X53918.1 | - | 293 | 63 | |||
More Information
Set Membership: |
|

WARS1
Genomic Map