Assay Details
| Gene Symbol: WARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174218.1 | NP_776643.1 | 2 - 3 | 500 | 68 | |
| GenBank mRNA | BC102806.1 | - | 3 - 4 | 445 | 68 | |
| X52113.1 | - | 427 | 68 | |||
| X53918.1 | - | 500 | 68 | |||
More Information
Set Membership: |
|

WARS1
Genomic Map