Assay Details
| Gene Symbol: WARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174218.1 | NP_776643.1 | 4 - 5 | 733 | 98 | |
| GenBank mRNA | BC102806.1 | - | 5 - 6 | 678 | 98 | |
| X52113.1 | - | 660 | 98 | |||
| X53918.1 | - | 733 | 98 | |||
More Information
Set Membership: |
|

WARS1
Genomic Map