Assay Details
| Gene Symbol: YARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174220.2 | NP_776645.1 | 5 - 6 | 684 | 145 | |
| GenBank mRNA | AF087021.1 | - | 5 - 6 | 684 | 145 | |
| BC122623.1 | - | 5 - 6 | 682 | 145 | ||
More Information
Set Membership: |
|

YARS1
Genomic Map