Assay Details
| Gene Symbol: ATP5PO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174244.1 | NP_776669.1 | 5 - 6 | 505 | 75 | |
| GenBank mRNA | AB099080.1 | - | 5 - 6 | 444 | 75 | |
| BC102204.1 | - | 5 - 6 | 466 | 75 | ||
| M18753.1 | - | 5 - 6 | 505 | 75 | ||
More Information
Set Membership: |
|

ATP5PO
Genomic Map