Assay Details
| Gene Symbol: BGLAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174249.2 | NP_776674.1 | 1 - 2 | 89 | 94 | |
| GenBank mRNA | BC126817.1 | - | 1 - 2 | 101 | 94 | |
| X51700.1 | - | 89 | 94 | |||
| X53699.1 | - | 89 | 94 | |||
More Information
Set Membership: |
|

BGLAP
Genomic Map