Assay Details
| Gene Symbol: CCNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174264.3 | NP_776689.2 | 5 - 6 | 737 | 63 | |
| GenBank mRNA | BC118382.1 | - | 5 - 6 | 644 | 63 | |
| BT020648.1 | - | 5 - 6 | 737 | 63 | ||
| BT020669.1 | - | 5 - 6 | 709 | 63 | ||
| BT020762.1 | - | 5 - 6 | 686 | 63 | ||
| BT020822.1 | - | 5 - 6 | 689 | 63 | ||
| BT021153.1 | - | 5 - 6 | 697 | 63 | ||
More Information
Set Membership: |
|

CCNB2
Genomic Map