Assay Details
| Gene Symbol: ACKR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174265.2 | NP_776690.1 | 1 | 1656 | 91 | |
| GenBank mRNA | BC142169.1 | - | 2 | 1263 | 91 | |
| BT020654.1 | - | 1 | 1190 | 91 | ||
| S63848.1 | - | 1 | 1656 | 91 | ||
More Information
Set Membership: |
|

ACKR4
Genomic Map