Assay Details
| Gene Symbol: CNMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174269.2 | NP_776694.1 | 2 - 3 | 241 | 135 | |
| GenBank mRNA | M65081.1 | - | 2 - 3 | 241 | 135 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNMD| Gene Symbol: CNMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174269.2 | NP_776694.1 | 2 - 3 | 241 | 135 | |
| GenBank mRNA | M65081.1 | - | 2 - 3 | 241 | 135 | |
Set Membership: |
|