Assay Details
| Gene Symbol: CRYGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174292.2 | NP_776717.1 | 1 - 2 | 32 | 70 | |
| GenBank mRNA | X03006.1 | - | 32 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174292.2, X03006.1 transcripts.
Set Membership: |
|

CRYGS
Genomic Map