Assay Details
| Gene Symbol: CRYGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174292.2 | NP_776717.1 | 2 - 3 | 270 | 143 | |
| GenBank mRNA | X03006.1 | - | 270 | 143 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRYGS| Gene Symbol: CRYGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174292.2 | NP_776717.1 | 2 - 3 | 270 | 143 | |
| GenBank mRNA | X03006.1 | - | 270 | 143 | ||
Set Membership: |
|