Assay Details
| Gene Symbol: CTLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174297.1 | NP_776722.1 | 1 - 2 | 101 | 74 | |
| GenBank mRNA | AB910936.1 | - | 1 - 2 | 101 | 74 | |
| AB910937.1 | - | 1 - 2 | 101 | 74 | ||
| AF539438.1 | - | 1 - 2 | 101 | 74 | ||
| LC271176.1 | - | 1 - 2 | 101 | 74 | ||
| X93305.1 | - | 101 | 74 | |||
More Information
Set Membership: |
|

CTLA4
Genomic Map