Assay Details
| Gene Symbol: CYP19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174305.1 | NP_776730.1 | 2 - 3 | 329 | 107 | |
| GenBank mRNA | DQ004741.1 | - | 1 - 2 | 224 | 107 | |
| DQ004742.1 | - | 1 - 2 | 224 | 107 | ||
| U18447.1 | - | 1 - 2 | 299 | 107 | ||
| Z32741.1 | - | 329 | 107 | |||
More Information
Set Membership: |
|

CYP19A1
Genomic Map