Assay Details
| Gene Symbol: NDUFA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174306.3 | NP_776731.1 | 3 - 4 | 274 | 70 | |
| GenBank mRNA | AJ011400.1 | - | 3 - 4 | 276 | 70 | |
| BC102502.1 | - | 1 | 278 | 70 | ||
More Information
Set Membership: |
|

NDUFA12
Genomic Map