Assay Details
| Gene Symbol: EPB42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174312.2 | NP_776737.1 | 6 - 7 | 982 | 57 | |
| GenBank mRNA | AF030029.1 | - | 6 - 7 | 982 | 57 | |
| AF030030.1 | - | 5 - 6 | 841 | 57 | ||
| BC151475.1 | - | 6 - 7 | 957 | 57 | ||
More Information
Set Membership: |
|

EPB42
Genomic Map