Assay Details
| Gene Symbol: GNAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174325.1 | NP_776750.1 | 8 - 9 | 1296 | 64 | |
| GenBank mRNA | BC142217.1 | - | 8 - 9 | 1316 | 64 | |
| M16116.1 | - | 8 - 9 | 1296 | 64 | ||
| S69263.1 | - | 1 - 2 | 33 | 64 | ||
| X12924.1 | - | 1259 | 64 | |||
More Information
Set Membership: |
|

GNAO1
Genomic Map