Assay Details
| Gene Symbol: HSD3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174343.3 | NP_776768.1 | 3 - 4 | 419 | 60 | |
| GenBank mRNA | BC111203.1 | - | 3 - 4 | 509 | 60 | |
| X17614.1 | - | 419 | 60 | |||
More Information
Set Membership: |
|

HSD3B1
Genomic Map