Assay Details
| Gene Symbol: ICAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174348.2 | NP_776773.1 | 2 - 3 | 883 | 84 | |
| GenBank mRNA | EU277736.1 | - | 2 - 3 | 330 | 84 | |
| U65789.1 | - | 2 - 3 | 883 | 84 | ||
More Information
Set Membership: |
|

ICAM1
Genomic Map