Assay Details
| Gene Symbol: ICAM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174349.1 | NP_776774.1 | 2 - 3 | 444 | 112 | |
| GenBank mRNA | L41844.1 | - | 2 - 3 | 444 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ICAM3| Gene Symbol: ICAM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174349.1 | NP_776774.1 | 2 - 3 | 444 | 112 | |
| GenBank mRNA | L41844.1 | - | 2 - 3 | 444 | 112 | |
Set Membership: |
|