Assay Details
| Gene Symbol: IL12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174355.2 | NP_776780.1 | 6 - 7 | 504 | 88 | |
| GenBank mRNA | EU276075.1 | - | 6 - 7 | 504 | 88 | |
| U14416.1 | - | 6 - 7 | 504 | 88 | ||
More Information
Set Membership: |
|

IL12A
Genomic Map