Assay Details
| Gene Symbol: IL12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174356.1 | NP_776781.1 | 1 - 2 | 89 | 66 | |
| GenBank mRNA | EU276076.1 | - | 1 - 2 | 89 | 66 | |
| U11815.1 | - | 1 - 2 | 89 | 66 | ||
More Information
Set Membership: |
|

IL12B
Genomic Map