Assay Details
| Gene Symbol: RPSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174379.2 | NP_776804.1 | 2 - 3 | 281 | 65 | |
| GenBank mRNA | AB099011.1 | - | 3 - 4 | 326 | 65 | |
| BC102490.1 | - | 3 - 4 | 331 | 65 | ||
| M64923.1 | - | 2 - 3 | 281 | 65 | ||
More Information
Set Membership: |
|

RPSA
Genomic Map