Assay Details
| Gene Symbol: RPSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174379.2 | NP_776804.1 | 3 - 4 | 532 | 59 | |
| GenBank mRNA | AB098943.1 | - | 1 - 2 | 92 | 59 | |
| BC102490.1 | - | 4 - 5 | 582 | 59 | ||
| JF979129.1 | - | 3 - 4 | 504 | 59 | ||
| M64923.1 | - | 3 - 4 | 532 | 59 | ||
More Information
Set Membership: |
|

RPSA
Genomic Map