Assay Details
| Gene Symbol: RPSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174379.2 | NP_776804.1 | 5 - 6 | 823 | 77 | |
| GenBank mRNA | AB098757.1 | - | 1 - 2 | 157 | 77 | |
| AB098775.1 | - | 1 - 2 | 209 | 77 | ||
| AB098943.1 | - | 3 - 4 | 386 | 78 | ||
| BC102490.1 | - | 6 - 7 | 873 | 77 | ||
| JF979129.1 | - | 5 - 6 | 795 | 77 | ||
| M64923.1 | - | 5 - 6 | 823 | 77 | ||
More Information
Set Membership: |
|

RPSA
Genomic Map