Assay Details
| Gene Symbol: MFAP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174386.2 | NP_776811.1 | 6 - 7 | 274 | 60 | |
| GenBank mRNA | BC102769.1 | - | 6 - 7 | 292 | 60 | |
| U37282.1 | - | 6 - 7 | 274 | 60 | ||
More Information
Set Membership: |
|

MFAP5
Genomic Map