Assay Details
| Gene Symbol: MMP13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174389.2 | NP_776814.1 | 6 - 7 | 919 | 68 | |
| GenBank mRNA | AF072685.1 | - | 6 - 7 | 919 | 68 | |
| AF135235.1 | - | 3 - 4 | 356 | 68 | ||
More Information
Set Membership: |
|

MMP13
Genomic Map