Assay Details
| Gene Symbol: MMP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174390.2 | NP_776815.1 | 8 - 9 | 1354 | 73 | |
| GenBank mRNA | AF290429.1 | - | 8 - 9 | 1354 | 73 | |
| BT026217.1 | - | 8 - 9 | 1473 | 73 | ||
More Information
Set Membership: |
|

MMP14
Genomic Map