Assay Details
| Gene Symbol: PPM1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174430.2 | NP_776855.1 | 2 - 3 | 930 | 87 | |
| GenBank mRNA | AJ005458.1 | - | 2 - 3 | 930 | 87 | |
| BC111235.1 | - | 2 - 3 | 1286 | 87 | ||
More Information
Set Membership: |
|

PPM1B
Genomic Map