Assay Details
| Gene Symbol: PROCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174437.1 | NP_776862.1 | 1 - 2 | 216 | 63 | |
| GenBank mRNA | BC116074.1 | - | 1 - 2 | 161 | 63 | |
| L39065.1 | - | 1 - 2 | 216 | 63 | ||
More Information
Set Membership: |
|

PROCR
Genomic Map