Assay Details
| Gene Symbol: PROCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174437.1 | NP_776862.1 | 2 - 3 | 469 | 83 | |
| GenBank mRNA | BC116074.1 | - | 2 - 3 | 414 | 83 | |
| L39065.1 | - | 2 - 3 | 469 | 83 | ||
More Information
Set Membership: |
|

PROCR
Genomic Map