Assay Details
| Gene Symbol: PROS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174438.1 | NP_776863.1 | 8 - 9 | 885 | 64 | |
| GenBank mRNA | M13044.1 | - | 8 - 9 | 885 | 64 | |
| X12891.1 | - | 885 | 64 | |||
More Information
Set Membership: |
|

PROS1
Genomic Map