Assay Details
| Gene Symbol: NMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174456.2 | NP_776881.1 | 8 - 9 | 996 | 62 | |
| GenBank mRNA | AF222687.1 | - | 8 - 9 | 996 | 62 | |
| AY007993.1 | - | 7 - 8 | 837 | 62 | ||
| BC112451.1 | - | 8 - 9 | 1035 | 62 | ||
More Information
Set Membership: |
|

NMT2
Genomic Map