Assay Details
| Gene Symbol: SPARC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174464.2 | NP_776889.1 | 9 - 10 | 942 | 70 | |
| GenBank mRNA | BC107529.1 | - | 9 - 10 | 961 | 70 | |
| BT030548.1 | - | 9 - 10 | 973 | 70 | ||
| J03233.1 | - | 9 - 10 | 942 | 70 | ||
More Information
Set Membership: |
|

SPARC
Genomic Map